Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap1g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF068707.1 | - | 2 | 319 | ||
AK052384.1 | - | 3 | 331 | |||
AK147177.1 | - | 3 | 358 | |||
AK151844.1 | - | 3 | 350 | |||
BC058402.1 | - | 2 | 441 | |||
BC080798.1 | - | 3 | 344 | |||
BC108374.1 | - | 3 | 345 | 4936738 | ||
BY013890.1 | - | 3 | 387 |