Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005165.2 | NP_005156.1 | 8 | 990 | ||
XM_005257949.2 | XP_005258006.1 | 9 | 1076 | |||
XM_011524556.1 | XP_011522858.1 | 9 | 1005 | |||
GenBank mRNA | AF054987.1 | - | 8 | 901 | ||
AK293453.1 | - | 8 | 857 | |||
AK293623.1 | - | 8 | 830 | |||
AK293693.1 | - | 7 | 835 | |||
AK294091.1 | - | 8 | 895 | |||
AK294157.1 | - | 8 | 883 | |||
AK296039.1 | - | 7 | 690 | |||
AK296053.1 | - | 8 | 1230 | |||
AK312281.1 | - | 8 | 894 | |||
BC003613.2 | - | 8 | 936 | 2987869 | ||
BC065565.1 | - | 8 | 936 | 6155379 | ||
BC103760.1 | - | 8 | 948 | |||
BC106925.1 | - | 8 | 901 | 40032967 | ||
BC106926.1 | - | 8 | 901 | 40032968 | ||
BT007006.1 | - | 7 | 845 | |||
CR541862.1 | - | 7 | 845 | |||
CR541881.1 | - | 7 | 845 |