Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007508.5 | NP_031534.2 | 4 | 463 | ||
GenBank mRNA | AK140873.1 | - | 4 | 439 | ||
AK149833.1 | - | 4 | 478 | |||
AK150742.1 | - | 4 | 449 | |||
AK153403.1 | - | 4 | 464 | |||
AK154869.1 | - | 4 | 464 | |||
AK160792.1 | - | 4 | 478 | |||
AK166857.1 | - | 4 | 453 | |||
AK170721.1 | - | 4 | 464 | |||
AK181657.1 | - | 2 | 288 | |||
AK216026.1 | - | 1 | 116 | |||
BC038392.1 | - | 4 | 441 | 2651677 |