Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC021653.1 | - | 12 | 2822 | ||
BC127082.1 | - | 12 | 2743 | 40111372 | ||
DQ086834.1 | - | 12 | 2782 | |||
L29006.1 | - | 11 | 3152 |