Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RND3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254738.1 | NP_001241667.1 | 5 | 2453 | ||
NM_005168.4 | NP_005159.1 | 6 | 2360 | |||
GenBank mRNA | BC012513.1 | - | 6 | 2317 | 4453777 | |
CF127383.1 | - | 1 | 151 | |||
X97758.1 | - | 5 | 2404 |