Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 8 | 1537 | ||
NM_007530.3 | NP_031556.1 | 7 | 1700 | |||
GenBank mRNA | AK090068.1 | - | 8 | 1514 | ||
AK144754.1 | - | 8 | 1511 | |||
AK205917.1 | - | 1 | 140 | |||
AK216433.1 | - | 1 | 135 | |||
BC003303.1 | - | 7 | 1604 | 3584161 | ||
BC021661.1 | - | 7 | 1711 | 2646883 | ||
BC040751.1 | - | 2 | 1890 |