Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Calm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007590.3 | NP_031616.1 | 6 | 2140 | ||
GenBank mRNA | AK049104.1 | - | 1 | 876 | ||
AK083996.1 | - | 3 | 1742 | |||
AK151610.1 | - | 6 | 2104 | |||
AK165568.1 | - | 6 | 2077 | |||
AK169630.1 | - | 6 | 2078 | |||
BC050926.1 | - | 6 | 2062 | 6411878 |