Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007648.4 | NP_031674.1 | 3, 5 | 152 | ||
GenBank mRNA | BC145926.1 | - | 3, 5 | 152 | 40131229 | |
BC145954.1 | - | 3, 5 | 152 | 40131257 | ||
EF474454.2 | - | 2, 4 | 74 | |||
GU456631.1 | - | 2, 4 | 74 | |||
J02990.1 | - | 3, 5 | 153 |