Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RAPGEF1 SIRNA Assays ›
Gene Symbol:
RAPGEF1
Gene Name:
Rap guanine nucleotide exchange factor 1
Gene Aliases:
C3G, GRF2
Chromosome Location:
Chr.9: 131576770 - 131740068 on Build GRCh38
UniGene ID:
Hs.127897
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
16016
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RAPGEF1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304275.1
NP_001291204.1
2
272
NM_005312.3
NP_005303.2
2
231
NM_198679.1
NP_941372.1
2
320
XM_005272186.4
XP_005272243.1
2
396
XM_005272191.3
XP_005272248.1
2
353
XM_006717067.3
XP_006717130.1
2
358
XM_006717072.3
XP_006717135.1
2
368
XM_006717074.3
XP_006717137.1
2
358
XM_011518569.2
XP_011516871.1
2
459
XM_011518570.2
XP_011516872.1
2
351
XM_011518571.2
XP_011516873.1
2
1019
XM_011518572.2
XP_011516874.1
2
198
XM_011518573.2
XP_011516875.1
2
331
XM_011518574.2
XP_011516876.1
2
351
XM_011518575.2
XP_011516877.1
2
351
XM_011518576.2
XP_011516878.1
2
351
XM_011518577.2
XP_011516879.1
2
351
XM_011518578.2
XP_011516880.1
2
351
XM_011518579.2
XP_011516881.1
2
351
XM_011518580.2
XP_011516882.1
2
351
XM_011518581.2
XP_011516883.1
2
351
XM_011518582.2
XP_011516884.1
2
351
XM_017014633.1
XP_016870122.1
2
368
XM_017014634.1
XP_016870123.1
2
195
XM_017014635.1
XP_016870124.1
2
249
XM_017014636.1
XP_016870125.1
2
244
XM_017014637.1
XP_016870126.1
2
351
XM_017014638.1
XP_016870127.1
2
351
XM_017014639.1
XP_016870128.1
2
351
XM_017014640.1
XP_016870129.1
2
351
XM_017014641.1
XP_016870130.1
2
351
GenBank mRNA
BC041710.1
-
2
199
6051007
BF058595.1
-
2
272
BG429001.1
-
2
321
CR749354.1
-
2
173
D21239.1
-
2
237
Back To Top
More Information
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPKK activity
blood vessel development
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of neuron projection development
single organismal cell-cell adhesion
Rap protein signal transduction
nerve growth factor signaling pathway
positive regulation of GTPase activity
negative regulation of Ras protein signal transduction
platelet-derived growth factor receptor signaling pathway
negative regulation of protein kinase B signaling
establishment of endothelial barrier
negative regulation of ERK1 and ERK2 cascade
cellular response to cAMP
negative regulation of canonical Wnt signaling pathway
regulation of cell junction assembly
cellular response to nerve growth factor stimulus
negative regulation of neural precursor cell proliferation
protein binding
Rap guanyl-nucleotide exchange factor activity
SH3 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0