Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007708.3 | NP_031734.3 | 3, 4 | 1137 | ||
GenBank mRNA | AF086823.1 | - | 3, 4 | 1193 | ||
AF086824.1 | - | 3, 4 | 1193 | |||
AK145037.1 | - | 4, 5 | 493 |