Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007723.2 | NP_031749.2 | 4 | 489 | ||
GenBank mRNA | BC085319.1 | - | 4 | 427 | ||
BC141261.1 | - | 4 | 258 | 9055873 | ||
BI738168.1 | - | 4 | 489 | |||
CX564336.1 | - | 4 | 470 | |||
M84742.1 | - | 4 | 261 | |||
U19717.1 | - | 3 | 224 |