Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007733.2 | NP_031759.2 | 38 | 2583 | ||
GenBank mRNA | AB000636.1 | - | 38 | 2557 | ||
AK019854.1 | - | 27 | 1689 | |||
AK149136.1 | - | 38 | 2576 | |||
BC118970.2 | - | 38 | 2536 | 40097873 |