Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 49, 50 | 3693 | ||
GenBank mRNA | AK075707.1 | - | 27, 28 | 2266 | ||
AK132496.1 | - | 49, 50 | 3689 | |||
AK159120.1 | - | 9, 10 | 894 | |||
AK159212.1 | - | 49, 50 | 3694 | |||
AK159408.1 | - | 49, 50 | 3690 | |||
AK159578.1 | - | 49, 50 | 3693 | |||
AK159958.1 | - | 28, 29 | 2313 | |||
AK160098.1 | - | 49, 50 | 3694 | |||
AK160787.1 | - | 49, 50 | 3689 | |||
AK160788.1 | - | 49, 50 | 3688 | |||
AK160790.1 | - | 49, 50 | 3689 | |||
AK160814.1 | - | 41, 42 | 3158 | |||
AK160946.1 | - | 49, 50 | 3692 | |||
AK161017.1 | - | 49, 50 | 3694 | |||
AK161605.1 | - | 49, 50 | 3689 | |||
AK164628.1 | - | 28, 29 | 2312 | |||
AK164808.1 | - | 49, 50 | 3689 | |||
BC007158.1 | - | 49, 50 | 3673 | 3487477 | ||
BC012438.1 | - | 10, 11 | 1133 | |||
BC042503.1 | - | 49, 50 | 3681 | 3586272 | ||
X58251.1 | - | 49, 50 | 3632 |