Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 50, 51 | 3876 | ||
GenBank mRNA | AK075707.1 | - | 28, 29 | 2449 | ||
AK132496.1 | - | 50, 51 | 3872 | |||
AK159120.1 | - | 10, 11 | 1076 | |||
AK159212.1 | - | 50, 51 | 3877 | |||
AK159408.1 | - | 50, 51 | 3873 | |||
AK159578.1 | - | 50, 51 | 3876 | |||
AK159958.1 | - | 29, 30 | 2496 | |||
AK160787.1 | - | 50, 51 | 3872 | |||
AK160788.1 | - | 50, 51 | 3871 | |||
AK160790.1 | - | 50, 51 | 3872 | |||
AK160814.1 | - | 42, 43 | 3341 | |||
AK160946.1 | - | 50, 51 | 3875 | |||
AK161017.1 | - | 50, 51 | 3877 | |||
AK161605.1 | - | 50, 51 | 3872 | |||
AK164628.1 | - | 29, 30 | 2495 | |||
AK164808.1 | - | 50, 51 | 3872 | |||
BC007158.1 | - | 50, 51 | 3856 | 3487477 | ||
BC012438.1 | - | 11, 12 | 1316 | |||
BC042503.1 | - | 50, 51 | 3864 | 3586272 | ||
X58251.1 | - | 50, 51 | 3815 |