Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007758.2 | NP_031784.1 | 2 | 345 | ||
GenBank mRNA | AK003072.2 | - | 2 | 345 | ||
BC109344.1 | - | 2 | 278 | 40054528 | ||
M35684.1 | - | 1 | 213 | |||
M61132.1 | - | 1 | 180 |