Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp3a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007819.4 | NP_031845.1 | 13 | 1791 | ||
GenBank mRNA | AK136494.1 | - | 13 | 1790 | ||
AK149572.1 | - | 13 | 1792 | |||
BC046592.1 | - | 13 | 1770 | 6308234 | ||
X63023.1 | - | 13 | 1767 |