Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007834.3 | NP_031860.1 | 2, 3 | 444 | ||
GenBank mRNA | AB001990.1 | - | 2, 3 | 259 | ||
AK077485.1 | - | 2, 3 | 258 | |||
AK077628.1 | - | 2, 3 | 250 | |||
AK188213.1 | - | 2, 3 | 240 | |||
AK214453.1 | - | 1, 2 | 157 | |||
AK216820.1 | - | 1, 2 | 100 | |||
BC003740.1 | - | 2, 3 | 232 | 3592608 | ||
BU709449.1 | - | 2, 3 | 646 |