Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gadd45a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007836.1 | NP_031862.1 | 4 | 664 | ||
GenBank mRNA | AK144900.1 | - | 4 | 664 | ||
BC011141.1 | - | 4 | 644 | 4035898 | ||
L28177.1 | - | 4 | 664 |