Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dsc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291809.1 | NP_001278738.1 | 3 | 471 | ||
NM_007882.3 | NP_031908.3 | 3 | 471 | |||
GenBank mRNA | BC144845.1 | - | 3 | 456 | ||
Y11169.1 | - | 3 | 471 |