Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK027977.1 | - | 5 | 823 | ||
BC004620.1 | - | 5 | 829 | 3585323 | ||
BC004703.1 | - | 5 | 1461 | 3499265 | ||
DV661455.1 | - | 4 | 425 | |||
X97755.1 | - | 5 | 827 |