Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110794.1 | NP_001104264.1 | 9 | 1100 | ||
NM_009674.3 | NP_033804.2 | 9 | 1215 | |||
GenBank mRNA | AK032013.1 | - | 9 | 991 | ||
AK077579.1 | - | 9 | 908 | |||
AK134589.1 | - | 10 | 985 | |||
AK149654.1 | - | 9 | 907 | |||
AK151438.1 | - | 9 | 909 | |||
AK161613.1 | - | 9 | 909 | |||
AK167588.1 | - | 9 | 883 | |||
AK169199.1 | - | 9 | 882 | |||
BC008997.1 | - | 9 | 899 | 3157926 | ||
L13129.1 | - | 9 | 902 |