Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009712.3 | NP_033842.3 | 2 | 516 | ||
GenBank mRNA | AK083309.1 | - | 2 | 500 | ||
AK140163.1 | - | 2 | 566 | |||
AK154098.1 | - | 2 | 436 | |||
AK158312.1 | - | 2 | 517 | |||
BC150661.1 | - | 2 | 473 | 9087572 |