Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001036684.2 | NP_001031761.1 | 20 | 3772 | ||
NM_009723.5 | NP_033853.1 | 22 | 3961 | |||
GenBank mRNA | AB030737.1 | - | 19 | 3333 | ||
AB030738.1 | - | 19 | 3333 | |||
AF053471.1 | - | 21 | 3790 | |||
AK054112.1 | - | 24 | 4059 | |||
AK147240.1 | - | 22 | 3847 | |||
AK147357.1 | - | 20 | 3701 | |||
AK147369.1 | - | 24 | 4195 | |||
AK147399.1 | - | 20 | 3924 | |||
AK147501.1 | - | 22 | 3908 | |||
GU734816.2 | - | 22 | 3468 |