Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pdia4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009787.2 | NP_033917.2 | 10 | 2043 | ||
GenBank mRNA | AK028292.1 | - | 10 | 1893 | ||
AK146288.1 | - | 10 | 1876 | |||
AK148801.1 | - | 10 | 1965 | |||
AK150566.1 | - | 10 | 1924 | |||
AK150903.1 | - | 2 | 551 | |||
AK161534.1 | - | 10 | 1881 | |||
AK168447.1 | - | 10 | 1847 | |||
AK169387.1 | - | 10 | 1888 | |||
BC066857.1 | - | 9 | 1642 | |||
BC141078.1 | - | 10 | 1939 | 9055690 | ||
J05186.1 | - | 10 | 1926 | |||
Y00884.1 | - | 9 | 1742 |