Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Capns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009795.3 | NP_033925.2 | 5, 6 | 459 | ||
GenBank mRNA | AF058298.1 | - | 4, 5 | 438 | ||
AK028092.1 | - | 3, 4 | 205 | |||
AK132819.1 | - | 4, 5 | 269 | |||
BC018352.1 | - | 5, 6 | 453 | 3156021 | ||
BC090988.1 | - | 5, 6 | 459 | 30246016 |