Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capzb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037761.2 | NP_001032850.1 | 6, 7 | 710 | ||
NM_001271405.1 | NP_001258334.1 | 6, 7 | 689 | |||
NM_001271406.1 | NP_001258335.1 | 6, 7 | 675 | |||
NM_009798.4 | NP_033928.1 | 6, 7 | 710 | |||
GenBank mRNA | AK153367.1 | - | 6, 7 | 487 | ||
AK153394.1 | - | 6, 7 | 487 | |||
AK156778.1 | - | 6, 7 | 679 | |||
AK159390.1 | - | 6, 7 | 684 | |||
AK160082.1 | - | 6, 7 | 685 | |||
AK162757.1 | - | 6, 7 | 677 | |||
AK168738.1 | - | 6, 7 | 638 | |||
AK177260.1 | - | 1, 2 | 38 | |||
AK177711.1 | - | 1, 2 | 37 | |||
AK179503.1 | - | 1, 2 | 38 | |||
AK182558.1 | - | 1, 2 | 38 | |||
AK182951.1 | - | 1, 2 | 38 | |||
AK186717.1 | - | 1, 2 | 38 | |||
AK187468.1 | - | 1, 2 | 38 | |||
AK187793.1 | - | 1, 2 | 38 | |||
AK188337.1 | - | 1, 2 | 38 | |||
AK190028.1 | - | 1, 2 | 38 | |||
AK192076.1 | - | 1, 2 | 38 | |||
AK198394.1 | - | 2, 3 | 128 | |||
AK200176.1 | - | 1, 2 | 38 | |||
AK207588.1 | - | 1, 2 | 38 | |||
AK212957.1 | - | 1, 2 | 101 | |||
BC002053.1 | - | 6, 7 | 652 | 3590980 | ||
FJ692320.1 | - | 6, 7 | 675 | |||
U10406.1 | - | 6, 7 | 630 | |||
U10407.1 | - | 6, 7 | 601 |