Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009802.2 | NP_033932.2 | 8 | 1061 | ||
GenBank mRNA | AF079834.1 | - | 7 | 1151 | ||
AF079835.1 | - | 8 | 1088 | |||
AK132599.1 | - | 8 | 1062 | |||
BC018582.1 | - | 8 | 1038 | |||
BC046495.1 | - | 7 | 837 | |||
BC049973.1 | - | 8 | 1059 | |||
BC055437.1 | - | 8 | 1047 | 4190566 |