Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009804.2 | NP_033934.2 | 7 | 969 | ||
GenBank mRNA | AK075853.1 | - | 7 | 939 | ||
AK088168.1 | - | 7 | 931 | |||
AK128919.1 | - | 7 | 944 | |||
AK133881.1 | - | 7 | 937 | |||
AK148961.1 | - | 7 | 937 | |||
AK150893.1 | - | 7 | 940 | |||
AK159152.1 | - | 7 | 940 | |||
AK159885.1 | - | 7 | 940 | |||
AK159891.1 | - | 7 | 891 | |||
AK159913.1 | - | 7 | 921 | |||
AK169069.1 | - | 7 | 934 | |||
AY040626.1 | - | 7 | 910 | |||
BC013447.1 | - | 7 | 926 | 4241333 | ||
BC047126.1 | - | 7 | 884 | |||
L25069.1 | - | 7 | 941 | |||
M62897.1 | - | 7 | 854 | |||
X52108.1 | - | 7 | 854 |