Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Runx1t1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111026.2 | NP_001104496.1 | 11 | 2523 | ||
NM_001111027.2 | NP_001104497.1 | 12 | 2717 | |||
NM_009822.3 | NP_033952.1 | 11 | 2455 | |||
GenBank mRNA | AK032132.1 | - | 11 | 2390 | ||
AK142006.1 | - | 11 | 2524 | |||
BC139200.1 | - | 11 | 2181 | 8862222 | ||
BC139201.1 | - | 11 | 2185 | 8862223 | ||
D32007.1 | - | 11 | 2455 |