Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd24a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009846.2 | NP_033976.1 | 2 | 713 | ||
GenBank mRNA | AK014279.1 | - | 2 | 713 | ||
AK133869.1 | - | 2 | 712 | |||
AK168111.1 | - | 2 | 712 | |||
AK169613.1 | - | 2 | 710 | |||
BC023197.1 | - | 1 | 437 | |||
BC026702.1 | - | 2 | 724 | |||
BC075622.1 | - | 2 | 693 | 6415159 | ||
M58661.1 | - | 2 | 701 | |||
X53825.1 | - | 2 | 651 |