Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161623.1 | NP_001155095.1 | 3 | 214 | ||
NM_009862.2 | NP_033992.2 | 4 | 453 | |||
GenBank mRNA | AF081536.1 | - | 4 | 453 | ||
AF081537.1 | - | 3 | 152 | |||
AF081538.1 | - | 3 | 152 | |||
AF098068.1 | - | 3 | 199 | |||
AJ223729.1 | - | 3 | 170 | |||
AK147013.1 | - | 3 | 170 | |||
AK167850.1 | - | 3 | 202 | |||
BB852042.1 | - | 3 | 166 | |||
BC006856.1 | - | 3 | 170 | |||
BC028635.1 | - | 3 | 206 | 1262927 |