Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdc45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009862.2 | NP_033992.2 | 4, 5 | 522 | ||
GenBank mRNA | AF081536.1 | - | 4, 5 | 522 | ||
AF081537.1 | - | 3, 4 | 221 | |||
AF081538.1 | - | 3, 4 | 221 | |||
AF098068.1 | - | 3, 4 | 268 | |||
AJ223729.1 | - | 3, 4 | 239 | |||
AK147013.1 | - | 3, 4 | 239 | |||
BC006856.1 | - | 3, 4 | 239 | |||
BC028635.1 | - | 3, 4 | 275 | 1262927 |