Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009868.4 | NP_033998.2 | 3 | 351 | ||
GenBank mRNA | AK164474.1 | - | 3 | 333 | ||
AK167620.1 | - | 3 | 283 | |||
BC054790.1 | - | 3 | 277 | 6831962 | ||
BY078746.1 | - | 3 | 353 | |||
D63942.1 | - | 3 | 312 | |||
X83930.2 | - | 3 | 266 |