Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Col4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009931.2 | NP_034061.2 | 52 | 5330 | ||
GenBank mRNA | AK146487.1 | - | 52 | 5279 | ||
AK147284.1 | - | 52 | 5252 | |||
AK147355.1 | - | 52 | 5255 | |||
AK147661.1 | - | 52 | 5296 | |||
AK159130.1 | - | 52 | 5292 | |||
BC002269.1 | - | 5 | 774 | |||
BC056620.1 | - | 11 | 1565 | |||
BC072650.1 | - | 53 | 5210 | 6852661 | ||
J04694.1 | - | 52 | 5234 | |||
X02201.1 | - | 10 | 1277 |