Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HINT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005340.6 | NP_005331.1 | 3 | 427 | ||
NR_024610.2 | 4 | 616 | ||||
NR_024611.2 | 3 | 462 | ||||
NR_073488.1 | 5 | 807 | ||||
NR_134494.1 | 5 | 772 | ||||
NR_134495.1 | 4 | 876 | ||||
GenBank mRNA | AK026557.1 | - | 3 | 336 | ||
AK054976.1 | - | 1 | 2419 | |||
AK056902.1 | - | 1 | 1758 | |||
BC001287.1 | - | 3 | 306 | 3458182 | ||
BC007090.1 | - | 3 | 379 | 4250172 | ||
BF967059.1 | - | 3 | 569 | |||
BG393782.1 | - | 5 | 666 | |||
BG527380.1 | - | 4 | 522 | |||
BG708765.1 | - | 3 | 432 | |||
CK002748.1 | - | 3 | 450 | |||
CN290233.1 | - | 3 | 279 | |||
CN290239.1 | - | 3 | 404 | |||
CR457048.1 | - | 3 | 284 | |||
DB125655.1 | - | 3 | 414 | |||
U27143.1 | - | 3 | 327 | |||
U51004.1 | - | 3 | 391 |