Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009948.2 | NP_034078.2 | 3, 4 | 373 | ||
GenBank mRNA | AB010826.1 | - | 2, 3 | 308 | ||
AF017174.2 | - | 3, 4 | 373 | |||
AK029141.1 | - | 3, 4 | 349 | |||
AK146851.1 | - | 3, 4 | 375 | |||
BC018270.1 | - | 3, 4 | 340 | 4920625 | ||
CJ092541.1 | - | 3, 4 | 373 |