Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEA11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011544.1 | NP_001011544.1 | 5 | 1107 | ||
NM_005366.4 | NP_005357.2 | 5 | 1106 | |||
XM_011531164.2 | XP_011529466.1 | 4 | 1737 | |||
XM_017029521.1 | XP_016885010.1 | 7 | 1317 | |||
XM_017029522.1 | XP_016885011.1 | 7 | 2701 | |||
GenBank mRNA | AK127849.1 | - | 7 | 2594 | ||
AY747607.1 | - | 5 | 1064 | |||
BC004479.2 | - | 5 | 1042 | 3833322 |