Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204084.2 | NP_001191013.1 | 4 | 1111 | ||
NM_001204085.1 | NP_001191014.1 | 4 | 1029 | |||
NM_001204086.2 | NP_001191015.1 | 4 | 1138 | |||
NM_001278164.1 | NP_001265093.1 | 4 | 1020 | |||
NM_001278165.1 | NP_001265094.1 | 4 | 1087 | |||
NM_001278166.1 | NP_001265095.1 | 4 | 931 | |||
NM_005380.7 | NP_005371.2 | 4 | 1144 | |||
NM_182744.3 | NP_877421.2 | 4 | 1039 | |||
GenBank mRNA | AK309257.1 | - | 4 | 1139 | ||
BC012037.1 | - | 4 | 942 | 3874204 | ||
D28124.1 | - | 4 | 911 |
Gene Symbol: MINOS1-NBL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204088.1 | NP_001191017.1 | 5 | 1188 | ||
NM_001204089.1 | NP_001191018.1 | 4 | 1003 | |||
GenBank mRNA | AK289456.1 | - | 4 | 1003 |