Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dync1i1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191023.1 | NP_001177952.1 | 6 | 826 | ||
NM_001191025.1 | NP_001177954.1 | 6 | 775 | |||
NM_001191026.1 | NP_001177955.1 | 7 | 802 | |||
NM_001191027.1 | NP_001177956.1 | 6 | 742 | |||
NM_010063.4 | NP_034193.2 | 7 | 835 | |||
GenBank mRNA | AF063229.1 | - | 7 | 669 | ||
AF063230.1 | - | 6 | 609 | |||
AK158589.1 | - | 7 | 803 | |||
BC051992.1 | - | 7 | 563 | 5696629 | ||
GU992206.1 | - | 6 | 567 | |||
GU992207.1 | - | 6 | 483 |