Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010073.2 | NP_034203.1 | 2 | 125 | ||
GenBank mRNA | AB013360.1 | - | 2 | 97 | ||
AK002468.1 | - | 2 | 109 | |||
AK004241.1 | - | 2 | 100 | |||
AK086634.1 | - | 1 | 401 | |||
AK179411.1 | - | 1 | 65 | |||
BC008256.1 | - | 2 | 125 | 2648752 |