Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST5 SIRNA Assays ›
Gene Symbol:
ST5
Gene Name:
suppression of tumorigenicity 5
Gene Aliases:
DENND2B, HTS1, p126
Chromosome Location:
Chr.11: 8693352 - 8910951 on Build GRCh38
UniGene ID:
Hs.117715
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
16161
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_005418.3
NP_005409.3
8
1900
NM_139157.2
NP_631896.1
4
368
NM_213618.1
NP_998783.1
5
1700
XM_005253077.1
XP_005253134.1
5
1615
XM_005253083.2
XP_005253140.1
4
510
XM_011520309.1
XP_011518611.1
5
1615
XM_011520310.1
XP_011518612.1
5
2027
XM_011520311.2
XP_011518613.1
5
2009
XM_011520312.1
XP_011518614.1
9
1974
XM_011520313.1
XP_011518615.1
8
1860
XM_011520314.1
XP_011518616.1
7
1752
XM_011520315.1
XP_011518617.1
7
1848
XM_011520316.1
XP_011518618.1
6
1778
XM_011520317.1
XP_011518619.1
5
1719
XM_011520318.1
XP_011518620.1
9
1974
XM_011520319.2
XP_011518621.1
5
1567
XM_011520320.1
XP_011518622.1
5
1615
XM_011520321.2
XP_011518623.1
5
1615
XM_011520322.1
XP_011518624.1
5
1615
XM_011520323.2
XP_011518625.2
4
438
XM_011520324.2
XP_011518626.2
4
438
XM_011520325.1
XP_011518627.1
4
510
XM_011520326.1
XP_011518628.1
8
714
XM_011520327.1
XP_011518629.1
4
403
XM_011520328.1
XP_011518630.1
4
435
XM_011520329.1
XP_011518631.1
8
714
XM_017018182.1
XP_016873671.1
6
1733
XM_017018183.1
XP_016873672.1
6
1710
XM_017018184.1
XP_016873673.1
8
1869
XM_017018185.1
XP_016873674.1
4
1665
XM_017018186.1
XP_016873675.1
4
1665
XM_017018187.1
XP_016873676.1
4
374
GenBank mRNA
AK127763.1
-
2
755
AK293245.1
-
2
133
AK302824.1
-
4
347
AK312305.1
-
5
1666
AK312758.1
-
4
710
BC036655.1
-
8
1905
4837231
BX647431.1
-
5
1700
S45936.1
-
1
38
U15131.1
-
5
1628
U15780.1
-
4
368
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of GTPase activity
Rab guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-lxknh:80/10.239.166.46:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0