Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013475.4 | NP_038503.4 | 5 | 517 | ||
GenBank mRNA | AA273732.1 | - | 2 | 132 | ||
AK010926.1 | - | 5 | 505 | |||
BC011037.1 | - | 5 | 533 | |||
BC019811.1 | - | 5 | 505 | 5125770 | ||
BC053338.1 | - | 5 | 493 | 5125963 | ||
D10056.1 | - | 5 | 491 | |||
S70439.1 | - | 5 | 501 | |||
Y11356.1 | - | 5 | 521 |