Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cryaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278569.1 | NP_001265498.1 | 1 | 165 | ||
NM_001278570.1 | NP_001265499.1 | 1 | 165 | |||
NM_013501.3 | NP_038529.1 | 1 | 165 | |||
GenBank mRNA | AJ310308.1 | - | 1 | 146 | ||
AK143463.1 | - | 1 | 164 | |||
BC085170.1 | - | 1 | 151 | 30699322 | ||
BC085172.1 | - | 1 | 151 | 30699371 | ||
BC092385.1 | - | 1 | 106 | 30602480 | ||
BY114969.1 | - | 1 | 163 | |||
EL604937.1 | - | 1 | 165 | |||
EL605017.1 | - | 1 | 167 | |||
J00376.1 | - | 1 | 65 |