Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243762.1 | NP_001230691.1 | 14 | 3080 | ||
NM_016691.4 | NP_057900.3 | 12 | 2698 | |||
GenBank mRNA | AK142227.1 | - | 12 | 2630 | ||
AK145763.1 | - | 12 | 2667 | |||
BC036347.1 | - | 12 | 2670 | 3989812 | ||
BU058610.1 | - | 1 | 35 |