Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 5, 6 | 720 | ||
NM_016878.4 | NP_058574.3 | 5, 6 | 544 | |||
GenBank mRNA | AF005051.1 | - | 4, 5 | 654 | ||
AK038646.1 | - | 720 | ||||
AK052001.1 | - | 522 | ||||
AK160084.1 | - | 5, 6 | 481 | |||
AK182360.1 | - | 2, 3 | 202 | |||
BC092232.1 | - | 5, 6 | 472 | 30551654 |