Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110831.1 | NP_001104301.1 | 6 | 758 | ||
NM_016878.4 | NP_058574.3 | 6 | 582 | |||
GenBank mRNA | AF005051.1 | - | 5 | 692 | ||
AK038646.1 | - | 758 | ||||
AK052001.1 | - | 560 | ||||
AK160084.1 | - | 6 | 519 | |||
AK182360.1 | - | 3 | 240 | |||
BC092232.1 | - | 6 | 510 | 30551654 |