Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v0a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243049.1 | NP_001229978.1 | 5, 6 | 500 | ||
NM_001243050.1 | NP_001229979.1 | 6, 7 | 678 | |||
NM_001243051.1 | NP_001229980.1 | 6, 7 | 699 | |||
NM_016920.3 | NP_058616.1 | 6, 7 | 678 | |||
GenBank mRNA | AB022321.1 | - | 5, 6 | 507 | ||
AF218249.1 | - | 6, 7 | 648 | |||
AF218250.1 | - | 5, 6 | 522 | |||
AF218251.1 | - | 5, 6 | 501 | |||
AK029316.1 | - | 6, 7 | 650 | |||
AK036530.1 | - | 6, 7 | 646 | |||
AK040343.1 | - | 6, 7 | 586 | |||
AK158786.1 | - | 5, 6 | 468 | |||
AK159114.1 | - | 6, 7 | 614 | |||
AK172078.1 | - | 6, 7 | 608 | |||
AK212043.1 | - | 1, 2 | 63 | |||
AW318708.1 | - | 6, 7 | 629 | |||
BC001995.1 | - | 6, 7 | 644 | |||
BC066839.1 | - | 6, 7 | 643 | 30101340 | ||
BC071182.1 | - | 6, 7 | 643 | 30101184 | ||
U13836.1 | - | 5, 6 | 540 |