Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap4s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021710.3 | NP_068356.1 | 6 | 920 | ||
GenBank mRNA | AF092093.1 | - | 6 | 849 | ||
AK005283.1 | - | 6 | 670 | |||
BC053339.1 | - | 6 | 680 | 5370721 | ||
BP426557.1 | - | 4 | 375 |