Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270996.1 | NP_001257925.1 | 2, 3 | 341 | ||
NM_007404.2 | NP_031430.2 | 2, 3 | 341 | |||
GenBank mRNA | AK122188.1 | - | 2, 3 | 324 | ||
AK148174.1 | - | 2, 3 | 227 | |||
AK155582.1 | - | 2, 3 | 302 | |||
AK155672.1 | - | 2, 3 | 234 | |||
AK161778.1 | - | 2, 3 | 265 | |||
BC047156.1 | - | 2, 3 | 219 | 6488707 | ||
BC052710.1 | - | 2, 3 | 213 | |||
BY780211.1 | - | 2, 3 | 285 | |||
CJ078757.1 | - | 2, 3 | 346 | |||
U41765.1 | - | 2, 3 | 213 |