Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009148.3 | NP_033174.2 | 2 | 225 | ||
GenBank mRNA | AF022962.1 | - | 2 | 251 | ||
AK010695.1 | - | 2 | 240 | |||
AK014499.1 | - | 2 | 248 | |||
AK036507.1 | - | 2 | 237 | |||
AK038125.1 | - | 2 | 248 | |||
AK039983.1 | - | 2 | 248 | |||
AK086603.1 | - | 2 | 244 | |||
AK173235.1 | - | 2 | 228 | |||
BC034644.1 | - | 2 | 225 | 5355276 |